Search
Search

4933402E13Rik| Gene Symbol: 4933402E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199996.1 | NP_001186925.1 | 4 | 402 | ||
| NM_001199997.1 | NP_001186926.1 | 4 | 363 | |||
| GenBank mRNA | AK016614.1 | - | 4 | 405 | ||
| BC132156.1 | - | 4 | 398 | 40130433 | ||