Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282012.1 | NP_001268941.1 | 3 | 612 | ||
NM_001282013.1 | NP_001268942.1 | 5 | 617 | |||
NM_001282024.1 | NP_001268953.1 | 4 | 711 | |||
NM_145487.3 | NP_663462.2 | 3 | 640 | |||
GenBank mRNA | AJ504719.1 | - | 3 | 1409 | ||
AK132945.1 | - | 3 | 340 | |||
AK203788.1 | - | 1 | 165 | |||
BC016636.1 | - | 3 | 561 | 2650633 | ||
BU148736.1 | - | 5 | 574 |