Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB089793.1 | - | 12 | 1154 | ||
AF482993.1 | - | 11 | 1118 | |||
AK049410.1 | - | 12 | 1150 | |||
AK051791.1 | - | 11 | 1057 | |||
AK129013.1 | - | 3 | 371 | |||
AK143281.1 | - | 12 | 1123 | |||
AK160826.1 | - | 10 | 992 | |||
AK173069.1 | - | 12 | 1490 | |||
BC030879.1 | - | 12 | 1153 | 4241988 | ||
BC034508.1 | - | 5 | 554 | |||
BC058947.1 | - | 12 | 1124 | 5702781 |