Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wibg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170869.2 | NP_001164340.1 | 3 | 302 | ||
NM_001253704.1 | NP_001240633.1 | 4 | 357 | |||
NM_001253705.1 | NP_001240634.1 | 3 | 461 | |||
NM_030100.4 | NP_084376.2 | 3 | 310 | |||
GenBank mRNA | AJ459407.1 | - | 3 | 227 | ||
AK020702.1 | - | 3 | 301 | |||
BC049647.1 | - | 3 | 235 | 6707378 | ||
BI989958.1 | - | 3 | 302 | |||
BQ177652.1 | - | 3 | 229 | |||
CK332572.1 | - | 3 | 251 |