Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289780.1 | NP_001276709.1 | 11 | 1456 | ||
NM_001289781.1 | NP_001276710.1 | 10 | 1789 | |||
NM_178403.5 | NP_848490.2 | 10 | 1438 | |||
GenBank mRNA | AK142271.1 | - | 10 | 1407 | ||
AK145380.1 | - | 10 | 1789 | |||
AK160849.1 | - | 10 | 1390 | |||
AK173290.1 | - | 10 | 1378 | |||
BC008544.1 | - | 10 | 1392 | 3497634 | ||
BC010600.1 | - | 2 | 169 | |||
BC138533.1 | - | 10 | 1280 | 8861553 | ||
BC145316.1 | - | 11 | 1298 | 9053871 |