Search
Search

N4bp1| Gene Symbol: N4bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030563.2 | NP_085040.2 | 4 | 2280 | ||
| GenBank mRNA | AK147522.1 | - | 4 | 2281 | ||
| AK172981.1 | - | 4 | 2262 | |||
| BC089324.1 | - | 3 | 1445 | |||
| BC111031.1 | - | 1 | 725 | |||