Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133189.3 | NP_573452.3 | 3 | 376 | ||
GenBank mRNA | AF361349.1 | - | 3 | 376 | ||
AF458899.1 | - | 3 | 376 | |||
AK032173.1 | - | 4 | 488 | |||
BC141277.1 | - | 3 | 414 | 9055889 |