Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CXCL2 SIRNA Assays ›
Gene Symbol:
Cxcl2
Gene Name:
chemokine (C-X-C motif) ligand 2
Gene Aliases:
Mip-2, Scyb2
Chromosome Location:
Chr.14: 18731346 - 18733391 on Build Rnor_6.0
UniGene ID:
Rn.10230
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS300112
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cxcl2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053647.1
NP_446099.1
2
198
GenBank mRNA
AB060092.1
-
2
198
S77604.1
-
2
153
U45965.1
-
2
161
X65647.1
-
2
138
Back To Top
More Information
Panther Classification:
Molecular Function -
chemokine
Gene Ontology Categories:
Function(s)
Process(es)
response to amphetamine
response to molecule of bacterial origin
positive regulation of leukocyte chemotaxis
inflammatory response
immune response
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
response to heat
response to gamma radiation
neutrophil chemotaxis
leukocyte chemotaxis
response to estradiol
wound healing
regulation of cell proliferation
response to glucocorticoid
chemokine-mediated signaling pathway
neuron death
cellular response to lipopolysaccharide
cellular response to interleukin-1
chemokine activity
CXCR chemokine receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-t8hgc:80/100.66.75.150:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0