Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN6 SIRNA Assays ›
Gene Symbol:
Ptpn6
Gene Name:
protein tyrosine phosphatase, non-receptor type 6
Gene Aliases:
Ptph6, Shp-1
Chromosome Location:
Chr.4: 157239141 - 157263890 on Build Rnor_6.0
UniGene ID:
Rn.18985
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS300586
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ptpn6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053908.1
NP_446360.1
3, 4
515
GenBank mRNA
BC099824.1
-
3, 4
372
7932620
U77038.1
-
3, 4
515
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
negative regulation of humoral immune response mediated by circulating immunoglobulin
protein dephosphorylation
cell proliferation
positive regulation of cell proliferation
response to wounding
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cell differentiation
platelet formation
abortive mitotic cell cycle
positive regulation of cell adhesion mediated by integrin
peptidyl-tyrosine dephosphorylation
intracellular signal transduction
megakaryocyte development
negative regulation of T cell proliferation
natural killer cell mediated cytotoxicity
negative regulation of MAP kinase activity
negative regulation of MAPK cascade
regulation of B cell differentiation
negative regulation of peptidyl-tyrosine phosphorylation
B cell receptor signaling pathway
negative regulation of B cell receptor signaling pathway
negative regulation of T cell receptor signaling pathway
regulation of release of sequestered calcium ion into cytosol
regulation of ERK1 and ERK2 cascade
platelet aggregation
regulation of G1/S transition of mitotic cell cycle
phosphotyrosine binding
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
SH3 domain binding
protein kinase binding
SH2 domain binding
natural killer cell lectin-like receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0