Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053921.1 | NP_446373.1 | 2, 3 | 451 | ||
XM_006246967.2 | XP_006247029.1 | 1, 2 | 237 | |||
XM_008767934.1 | XP_008766156.1 | 1, 2 | 242 | |||
GenBank mRNA | AB002111.1 | - | 2, 3 | 451 | ||
BC072481.1 | - | 2, 3 | 407 | 7104469 |
Gene Symbol: LOC100909787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750890.3 | XP_003750938.1 | 2, 3 | 451 | ||
XM_006247026.2 | XP_006247088.1 | 1, 2 | 237 | |||
XM_008773868.1 | XP_008772090.1 | 1, 2 | 242 |