siRNA Details
Gene Symbol: P2ry6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057124.2 | NP_476465.1 | 3 | 531 | ||
XM_006229711.2 | XP_006229773.1 | 2 | 615 | |||
XM_006229712.2 | XP_006229774.1 | 3 | 1967 | |||
XM_006229713.2 | XP_006229775.1 | 3 | 684 | |||
XM_006229714.2 | XP_006229776.1 | 3 | 1631 | |||
XM_006229715.2 | XP_006229777.1 | 4 | 2114 | |||
XM_006229716.2 | XP_006229778.1 | 3 | 1352 | |||
XM_006229717.2 | XP_006229779.1 | 3 | 3922 | |||
GenBank mRNA | BC072520.1 | - | 3 | 531 | 7105972 | |
D63665.1 | - | 4 | 755 |
