Search
Search

Sv2a| Gene Symbol: Sv2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057210.2 | NP_476558.2 | 10 | 1794 | ||
| XM_006232952.2 | XP_006233014.1 | 11 | 2223 | |||
| GenBank mRNA | BC092132.1 | - | 10 | 1794 | 7314271 | |
| L05435.1 | - | 11 | 2140 | |||