Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133412.1 | NP_596903.1 | 3 | 897 | ||
XM_006245576.2 | XP_006245638.1 | 3 | 967 | |||
GenBank mRNA | AF321415.1 | - | 3 | 897 |
Gene Symbol: LOC102554121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245586.2 | XP_006245648.1 | 3 | 968 |