Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cblb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133601.1 | NP_598285.1 | 4 | 476 | ||
XM_006248258.2 | XP_006248320.1 | 4 | 550 | |||
XM_006248259.2 | XP_006248321.1 | 4 | 633 | |||
XM_006248260.2 | XP_006248322.1 | 4 | 549 | |||
XM_008768626.1 | XP_008766848.1 | 3 | 451 | |||
XM_008768627.1 | XP_008766849.1 | 4 | 746 | |||
GenBank mRNA | AB071283.1 | - | 4 | 476 |