Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134413.2 | NP_599240.1 | 5, 6 | 1416 | ||
XM_006255214.2 | XP_006255276.1 | 5, 6 | 1410 | |||
XM_006255215.2 | XP_006255277.1 | 5, 6 | 1330 | |||
XM_008772346.1 | XP_008770568.1 | 5, 6 | 1433 | |||
GenBank mRNA | AF015911.1 | - | 6, 7 | 1417 |