Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042354.1 | NP_001035813.1 | 2, 3 | 232 | ||
NM_001042356.1 | NP_001035815.1 | 2, 3 | 232 | |||
NM_021739.2 | NP_068507.2 | 2, 3 | 232 | |||
XM_006251170.2 | XP_006251232.1 | 2, 3 | 331 | |||
XM_006251171.2 | XP_006251233.1 | 2, 3 | 331 | |||
XM_006251172.2 | XP_006251234.1 | 2, 3 | 331 | |||
XM_006251174.2 | XP_006251236.1 | 2, 3 | 331 | |||
XM_006251176.2 | XP_006251238.1 | 2, 3 | 312 | |||
XM_006251177.2 | XP_006251239.1 | 2, 3 | 311 | |||
GenBank mRNA | CB616255.1 | - | 2, 3 | 232 | ||
M16112.1 | - | 2, 3 | 212 | |||
X83375.1 | - | 2, 3 | 194 |