Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012570.2 | NP_036702.1 | 8, 9 | 1357 | ||
GenBank mRNA | BC081841.1 | - | 8, 9 | 1241 | 7107138 | |
X14044.1 | - | 8, 9 | 1247 | |||
X14223.1 | - | 8, 9 | 1220 |