Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADIPOQ SIRNA Assays ›
Gene Symbol:
Adipoq
Gene Name:
adiponectin, C1Q and collagen domain containing
Gene Aliases:
Acdc, Acrp30
Chromosome Location:
Chr.11: 81330845 - 81344488 on Build Rnor_6.0
UniGene ID:
Rn.24299
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS302505
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Adipoq
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_144744.3
NP_653345.1
3
480
GenBank mRNA
AY033885.1
-
3
427
BC092565.1
-
2
416
7308618
FM047633.1
-
3
480
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009605, AK003138, AX767957, BC028770, AY754346, and others.
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
positive regulation of protein phosphorylation
glucose metabolic process
fatty acid beta-oxidation
response to nutrient
circadian rhythm
response to sucrose
response to glucose
positive regulation of signal transduction
negative regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of protein kinase A signaling
negative regulation of macrophage derived foam cell differentiation
negative regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of cholesterol efflux
regulation of glucose metabolic process
response to activity
negative regulation of smooth muscle cell migration
fatty acid oxidation
negative regulation of cell migration
negative regulation of granulocyte differentiation
response to nutrient levels
negative regulation of protein autophosphorylation
positive regulation of cellular protein metabolic process
negative regulation of tumor necrosis factor production
positive regulation of interleukin-8 production
cellular response to insulin stimulus
positive regulation of myeloid cell apoptotic process
adiponectin-activated signaling pathway
negative regulation of heterotypic cell-cell adhesion
low-density lipoprotein particle clearance
response to tumor necrosis factor
cellular response to drug
response to drug
glucose homeostasis
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of MAP kinase activity
response to ethanol
negative regulation of fat cell differentiation
negative regulation of macrophage differentiation
negative regulation of low-density lipoprotein particle receptor biosynthetic process
negative regulation of gluconeogenesis
negative regulation of blood pressure
positive regulation of blood pressure
positive regulation of protein kinase activity
negative regulation of transcription, DNA-templated
positive regulation of fatty acid metabolic process
positive regulation of glucose import
negative regulation of hormone secretion
negative regulation of smooth muscle cell proliferation
negative regulation of inflammatory response
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of phagocytosis
negative regulation of synaptic transmission
brown fat cell differentiation
protein homooligomerization
response to glucocorticoid
membrane depolarization
membrane hyperpolarization
protein heterotrimerization
negative regulation of ERK1 and ERK2 cascade
response to linoleic acid
detection of oxidative stress
cellular response to cAMP
positive regulation of monocyte chemotactic protein-1 production
cellular response to epinephrine stimulus
protein localization to plasma membrane
negative regulation of intracellular protein transport
negative regulation of receptor binding
negative regulation of DNA biosynthetic process
positive regulation of glycogen (starch) synthase activity
positive regulation of metanephric glomerular visceral epithelial cell development
positive regulation of cAMP-dependent protein kinase activity
positive regulation of renal albumin absorption
negative regulation of platelet-derived growth factor receptor-alpha signaling pathway
negative regulation of metanephric mesenchymal cell migration
receptor binding
hormone activity
sialic acid binding
identical protein binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0