Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104633.1 | NP_001098103.1 | 15 | 2148 | ||
GenBank mRNA | AB288378.1 | - | 14 | 1613 | ||
AF268594.1 | - | 8 | 838 |