Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other JAK3 SIRNA Assays ›
Gene Symbol:
Jak3
Gene Name:
Janus kinase 3
Gene Aliases:
RATJAK3
Chromosome Location:
Chr.16: 20107468 - 20120678 on Build Rnor_6.0
UniGene ID:
Rn.11159
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS303438
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Jak3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012855.2
NP_036987.2
10
1431
XM_008771064.1
XP_008769286.1
10
2529
GenBank mRNA
D28508.1
-
10
1431
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of dendritic cell cytokine production
inflammatory response
enzyme linked receptor protein signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of cytosolic calcium ion concentration
cell migration
peptidyl-tyrosine phosphorylation
cytokine-mediated signaling pathway
B cell differentiation
erythrocyte differentiation
negative regulation of interleukin-10 production
negative regulation of interleukin-12 production
intracellular signal transduction
interleukin-4-mediated signaling pathway
peptidyl-tyrosine autophosphorylation
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
tyrosine phosphorylation of Stat5 protein
regulation of apoptotic process
T cell homeostasis
innate immune response
negative regulation of FasL biosynthetic process
negative regulation of T-helper 1 cell differentiation
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
positive regulation of immune response
negative regulation of T cell activation
positive regulation of calcium ion transport
regulation of T cell apoptotic process
negative regulation of thymocyte apoptotic process
response to interleukin-4
positive regulation of dendritic cell apoptotic process
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
ATP binding
transcription factor binding
protein phosphatase binding
hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8bdhq:80/100.66.78.62:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0