Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sclt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153740.1 | NP_714962.1 | 7 | 475 | ||
XM_006232296.2 | XP_006232358.1 | 7 | 937 | |||
XM_008760906.1 | XP_008759128.1 | 3 | 179 | |||
GenBank mRNA | AF421190.1 | - | 7 | 475 | ||
AF421191.1 | - | 7 | 470 | |||
AF427094.1 | - | 7 | 475 | |||
AF427095.1 | - | 6 | 424 |