Search
Search

Fam149b1| Gene Symbol: Fam149b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013878.1 | NP_001013900.1 | 10 | 1315 | ||
| XM_006251632.2 | XP_006251694.1 | 11 | 1586 | |||
| XM_006251634.2 | XP_006251696.1 | 9 | 1216 | |||
| GenBank mRNA | BC087126.1 | - | 10 | 1315 | 7111155 | |