Search
Search

Katnb1| Gene Symbol: Katnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024746.1 | NP_001019917.1 | 7 | 533 | ||
| XM_006255078.1 | XP_006255140.1 | 8 | 909 | |||
| XM_008772277.1 | XP_008770499.1 | 8 | 850 | |||
| XM_008772278.1 | XP_008770500.1 | 8 | 981 | |||
| XM_008772279.1 | XP_008770501.1 | 8 | 1032 | |||
| XM_008772280.1 | XP_008770502.1 | 8 | 910 | |||
| GenBank mRNA | AY953248.1 | - | 7 | 533 | ||
| BC161823.1 | - | 7 | 779 | |||