Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207591.1 | NP_997474.1 | 4 | 448 | ||
GenBank mRNA | AY535002.1 | - | 4 | 448 | ||
BC086359.1 | - | 4 | 452 | 7304013 | ||
FQ212888.1 | - | 4 | 479 | |||
FQ213201.1 | - | 4 | 457 | |||
FQ218937.1 | - | 4 | 454 | |||
FQ219058.1 | - | 4 | 454 | |||
FQ222789.1 | - | 4 | 458 |