Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134972.1 | NP_001128444.1 | 15, 16 | 2205 | ||
XM_006228613.2 | XP_006228675.1 | 13, 14 | 2051 | |||
XM_006228614.1 | XP_006228676.1 | 14, 15 | 1958 | |||
XM_006228615.2 | XP_006228677.1 | 15, 16 | 2894 | |||
XM_006228616.2 | XP_006228678.1 | 15, 16 | 1647 | |||
XM_006228617.2 | XP_006228679.1 | 14, 15 | 1694 | |||
XM_006228618.2 | XP_006228680.1 | 14, 15 | 1691 | |||
XM_006228619.2 | XP_006228681.1 | 14, 15 | 1649 | |||
XM_006228620.2 | XP_006228682.1 | 14, 15 | 1646 |