Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911519 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749011.3 | XP_003749059.1 | 14 | 3513 | ||
XM_006230592.2 | XP_006230654.1 | 14 | 3063 | |||
XM_006230593.2 | XP_006230655.1 | 13 | 3537 | |||
XM_008760030.1 | XP_008758252.1 | 13 | 3585 | |||
XM_008760031.1 | XP_008758253.1 | 14 | 2969 |
Gene Symbol: Pidd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106318.2 | NP_001099788.2 | 14 | 2440 | ||
XM_006230543.2 | XP_006230605.1 | 14 | 3063 | |||
XM_006230544.2 | XP_006230606.1 | 13 | 3537 | |||
XM_008759983.1 | XP_008758205.1 | 13 | 3585 | |||
XM_008759984.1 | XP_008758206.1 | 14 | 2969 |