Search
Search

Kpna5| Gene Symbol: Kpna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025113.1 | NP_001020284.1 | 10, 11 | 1123 | ||
| XM_006256442.2 | XP_006256504.1 | 11, 12 | 1303 | |||
| XM_008772879.1 | XP_008771101.1 | 9, 10 | 978 | |||
| GenBank mRNA | AY779029.1 | - | 10, 11 | 1123 | ||