Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERBB3 SIRNA Assays ›
Gene Symbol:
Erbb3
Gene Name:
erb-b2 receptor tyrosine kinase 3
Gene Aliases:
nuc-ErbB3
Chromosome Location:
Chr.7: 2991016 - 3010312 on Build Rnor_6.0
UniGene ID:
Rn.10228
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS309602
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Erbb3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017218.3
NP_058914.2
19
2376
GenBank mRNA
U29339.2
-
19
2376
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
endocardial cushion development
negative regulation of cell adhesion
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
epidermal growth factor receptor signaling pathway
axonogenesis
peripheral nervous system development
skeletal muscle tissue development
circadian rhythm
response to wounding
negative regulation of signal transduction
Schwann cell differentiation
phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cranial nerve development
signal transduction by protein phosphorylation
cellular response to insulin stimulus
regulation of cell proliferation
odontogenesis
response to drug
negative regulation of neuron apoptotic process
tongue development
positive regulation of glucose import
negative regulation of secretion
neuron apoptotic process
mammary gland involution
positive regulation of protein tyrosine kinase activity
positive regulation of calcineurin-NFAT signaling cascade
extrinsic apoptotic signaling pathway in absence of ligand
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
ATP binding
growth factor binding
protein tyrosine kinase activator activity
neuregulin receptor activity
neuregulin binding
identical protein binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0