Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1560175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248356.2 | XP_006248418.1 | 7 | 1116 | ||
XM_006248357.2 | XP_006248419.1 | 6 | 1000 | |||
XM_221454.8 | XP_221454.5 | 7 | 1251 |