Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pds5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083624.1 | NP_001077093.1 | 20 | 2339 | ||
XM_008770153.1 | XP_008768375.1 | 20 | 2373 | |||
XM_008770154.1 | XP_008768376.1 | 19 | 2286 | |||
GenBank mRNA | EF460313.1 | - | 20 | 2339 |