Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191680.1 | NP_001178609.1 | 6, 7 | 813 | ||
XM_006251243.2 | XP_006251305.1 | 7, 8 | 1115 | |||
XM_006251244.2 | XP_006251306.1 | 7, 8 | 1112 | |||
XM_006251245.2 | XP_006251307.1 | 7, 8 | 948 | |||
XM_006251246.2 | XP_006251308.1 | 7, 8 | 945 |