Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191682.1 | NP_001178611.1 | 17, 18 | 1864 | ||
XM_006251285.2 | XP_006251347.1 | 18, 19 | 2263 | |||
XM_006251287.2 | XP_006251349.1 | 19, 20 | 2319 | |||
XM_006251288.2 | XP_006251350.1 | 18, 19 | 2263 | |||
XM_006251289.2 | XP_006251351.1 | 18, 19 | 2263 | |||
XM_008770294.1 | XP_008768516.1 | 17, 18 | 2360 | |||
XM_008770295.1 | XP_008768517.1 | 12, 13 | 2146 |