Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sacs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252167.2 | XP_006252229.1 | 11 | 5260 | ||
XM_006252168.2 | XP_006252230.1 | 10 | 4788 | |||
XM_006252169.2 | XP_006252231.1 | 10 | 5233 | |||
XM_008770792.1 | XP_008769014.1 | 10 | 5113 |