Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173527.1 | NP_001166998.1 | 1 | 625 | ||
GenBank mRNA | BC098730.1 | - | 1 | 625 |