Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107861.1 | NP_001101331.1 | 4 | 2036 | ||
XM_006236547.2 | XP_006236609.1 | 4 | 2347 | |||
XM_006236548.2 | XP_006236610.1 | 4 | 2191 | |||
XM_006236549.2 | XP_006236611.1 | 4 | 2608 | |||
XM_008762892.1 | XP_008761114.1 | 4 | 2713 | |||
XM_008762893.1 | XP_008761115.1 | 5 | 2306 | |||
XM_008762894.1 | XP_008761116.1 | 4 | 2081 |