Search
Search

Zfp638| Gene Symbol: Zfp638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107868.1 | NP_001101338.1 | 22 | 4364 | ||
| XM_006236749.2 | XP_006236811.1 | 22 | 4507 | |||
| XM_006236750.2 | XP_006236812.1 | 21 | 4438 | |||
| XM_006236751.2 | XP_006236813.1 | 22 | 4507 | |||
| XM_006236753.2 | XP_006236815.1 | 20 | 4354 | |||
| XM_006236754.2 | XP_006236816.1 | 22 | 4526 | |||