Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Murc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107931.2 | NP_001101401.1 | 2 | 440 | ||
GenBank mRNA | EU487255.1 | - | 2 | 440 |