siRNA Details
Gene Symbol: Dmrta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107945.1 | NP_001101415.1 | 2 | 1333 |

Gene Symbol: Dmrta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107945.1 | NP_001101415.1 | 2 | 1333 |