Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238423.2 | XP_006238485.1 | 2 | 138 | ||
XM_006238424.2 | XP_006238486.1 | 2 | 144 | |||
XM_008763915.1 | XP_008762137.1 | 2 | 138 |