Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SKI SIRNA Assays ›
Gene Symbol:
Ski
Gene Name:
SKI proto-oncogene
Gene Aliases:
RGD1565591, c-ski
Chromosome Location:
Chr.5: 172556196 - 172624204 on Build Rnor_6.0
UniGene ID:
Rn.233489
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS320452
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ski
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_008764420.1
XP_008762642.1
6, 7
1983
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011385, BC030841, BC068305, CB722801, AK019148.
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neural tube closure
lens morphogenesis in camera-type eye
transcription, DNA-templated
negative regulation of cell proliferation
anterior/posterior axis specification
negative regulation of Schwann cell proliferation
myotube differentiation
olfactory bulb development
myelination in peripheral nervous system
embryonic limb morphogenesis
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
negative regulation of histone deacetylation
negative regulation of activin receptor signaling pathway
somatic stem cell population maintenance
regulation of apoptotic process
camera-type eye development
negative regulation of catalytic activity
positive regulation of DNA binding
nose morphogenesis
negative regulation of osteoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of fibroblast proliferation
camera-type eye morphogenesis
skeletal muscle fiber development
palate development
retina development in camera-type eye
face morphogenesis
bone morphogenesis
SMAD protein signal transduction
protein homotrimerization
chromatin binding
transcription corepressor activity
zinc ion binding
protein kinase binding
protein domain specific binding
ubiquitin protein ligase binding
SMAD binding
histone deacetylase inhibitor activity
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0