Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191770.1 | NP_001178699.1 | 9 | 1764 | ||
XM_008764459.1 | XP_008762681.1 | 10 | 2193 | |||
XM_008764460.1 | XP_008762682.1 | 10 | 1901 | |||
XM_008764461.1 | XP_008762683.1 | 10 | 1893 | |||
XM_008764462.1 | XP_008762684.1 | 9 | 2272 | |||
XM_008764463.1 | XP_008762685.1 | 8 | 1698 |