Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766067.1 | XP_008764289.1 | 13 | 1917 | ||
XM_008766068.1 | XP_008764290.1 | 12 | 1902 |