siRNA Details
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153624.2 | NP_705888.2 | 8 | 1212 | ||
XM_006254403.2 | XP_006254465.1 | 8 | 1268 |

Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153624.2 | NP_705888.2 | 8 | 1212 | ||
XM_006254403.2 | XP_006254465.1 | 8 | 1268 |