Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759936.1 | XP_008758158.1 | 8 | 2265 |
Gene Symbol: LOC100911588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759962.1 | XP_008758184.1 | 3 | 394 |