Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760646.1 | XP_008758868.1 | 15 | 1105 | ||
XM_008760647.1 | XP_008758869.1 | 15 | 1102 | |||
XM_008760648.1 | XP_008758870.1 | 15 | 1080 | |||
XM_008760649.1 | XP_008758871.1 | 15 | 1077 | |||
XM_008760650.1 | XP_008758872.1 | 14 | 1015 | |||
XM_008760651.1 | XP_008758873.1 | 14 | 1011 | |||
XM_008760652.1 | XP_008758874.1 | 14 | 990 | |||
XM_008760653.1 | XP_008758875.1 | 14 | 986 | |||
GenBank mRNA | FQ221531.1 | - | 15 | 977 |