Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108545.1 | NP_001102015.1 | 3 | 828 | ||
XM_006232087.2 | XP_006232149.1 | 2 | 2851 | |||
XM_008760775.1 | XP_008758997.1 | 3 | 620 | |||
XM_008760776.1 | XP_008758998.1 | 4 | 524 |