siRNA Details
Gene Symbol: Rnf115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108560.1 | NP_001102030.1 | 4 | 376 | ||
XM_006232975.1 | XP_006233037.1 | 4 | 397 |

Gene Symbol: Rnf115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108560.1 | NP_001102030.1 | 4 | 376 | ||
XM_006232975.1 | XP_006233037.1 | 4 | 397 |