Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam76a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108686.1 | NP_001102156.1 | 8 | 965 | ||
GenBank mRNA | FQ211538.1 | - | 6 | 898 |