Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022236.1 | NP_071572.1 | 14, 15 | 1697 | ||
GenBank mRNA | AB027155.1 | - | 14, 15 | 1697 | ||
AB027156.1 | - | 2032 | ||||
AY462091.1 | - | 1663 | ||||
AY462092.1 | - | 1771 | ||||
AY462093.1 | - | 1809 | ||||
AY462094.1 | - | 1478 | ||||
AY462095.1 | - | 1944 |