Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022399.2 | NP_071794.1 | 8 | 1063 | ||
GenBank mRNA | BC062395.1 | - | 8 | 1040 | 5623178 | |
D78308.1 | - | 8 | 1015 | |||
FQ210587.1 | - | 1068 | ||||
FQ226996.1 | - | 2 | 148 | |||
X13702.1 | - | 2 | 196 | |||
X53363.1 | - | 8 | 1035 | |||
X79327.1 | - | 8 | 1013 |